Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31554320UL
Description
COG1 Polyclonal antibody specifically detects COG1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| COG1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| COG complex subunit 1, component of oligomeric golgi complex 1CDG2G, conserved oligomeric Golgi complex subunit 1, KIAA1381DKFZp762L1710, LDLBconserved oligomeric Golgi complex protein 1, low density lipoprotein receptor defect B complementing | |
| A synthetic peptide corresponding to a sequence within amino acids 900-993 of human COG1 (NP_061184.1). | |
| 20 μg | |
| Signal Transduction | |
| 9382 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction