Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$148.20 - $378.00
Specifications
Antigen | COG1 |
---|---|
Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
COG1 Polyclonal antibody specifically detects COG1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
COG1 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS, 50% glycerol, pH7.3 | |
9382 | |
IgG | |
Affinity purified |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
COG complex subunit 1, component of oligomeric golgi complex 1CDG2G, conserved oligomeric Golgi complex subunit 1, KIAA1381DKFZp762L1710, LDLBconserved oligomeric Golgi complex protein 1, low density lipoprotein receptor defect B complementing | |
A synthetic peptide corresponding to a sequence within amino acids 900-993 of human COG1 (NP_061184.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title