Learn More
Description
Specifications
Specifications
| Antigen | COG2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000 |
| Formulation | PBS and 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. | Q14746 |
| Gene Alias | component of oligomeric golgi complex 2COG complex subunit 2, conserved oligomeric Golgi complex protein 2, conserved oligomeric Golgi complex subunit 2, LDLCbrefeldin A-sensitive, peripheral Golgi protein, low density lipoprotein receptor defect C complementing, Low density lipoprotein receptor defect C-complementing protein |
| Gene Symbols | COG2 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
