Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coilin Rabbit anti-Human, Mouse, Clone: 7Z4K1, Novus Biologicals™
SDP

Catalog No. NB167483 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167483 100 μg
NB167484 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167483 Supplier Novus Biologicals Supplier No. NBP316196100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Coilin Monoclonal antibody specifically detects Coilin in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen Coilin
Applications Western Blot
Classification Monoclonal
Clone 7Z4K1
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CLN80p80-coilin, coilin, coilin p80, p80
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Coilin (P38432).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 8161
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.