Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coilin Rabbit anti-Human, Mouse, Clone: 7Z4K1, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Coilin |
---|---|
Clone | 7Z4K1 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Coilin Monoclonal antibody specifically detects Coilin in Human, Mouse samples. It is validated for Western BlotSpecifications
Coilin | |
Western Blot | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
8161 | |
IgG | |
Affinity purified |
7Z4K1 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
CLN80p80-coilin, coilin, coilin p80, p80 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Coilin (P38432). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title