Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COL4A3BP Antibody, Novus Biologicals™
SDP

Catalog No. NB1002114 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002114 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002114 Supplier Novus Biologicals Supplier No. NB1002114
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

COL4A3BP Polyclonal specifically detects COL4A3BP in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen COL4A3BP
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2500-1:10000, Immunoprecipitation 1-4 ug/mg of lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. Q9Y5P4
Gene Alias Ceramide transfer protein, CERTFLJ20597, CERTL, collagen type IV alpha-3-binding protein, collagen, type IV, alpha 3 (Goodpasture antigen) binding protein, Goodpasture antigen-binding protein, GPBPSTARD11ceramide transporter, hCERT, lipid-transfer protein CERTL, StARD11, StAR-related lipid transfer (START) domain containing 11, StAR-related lipid transfer protein 11, START domain containing 11, START domain-containing protein 11
Gene Symbols COL4A3BP
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 300 and 350 of human CERTL(GPBP) using the numbering given in entry NP_005704.1 (GeneID 10087).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers, Immune System Diseases, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10087
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.