Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COL4A3BP Antibody, Novus Biologicals™
SDP

Catalog No. NB10093386 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093386 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10093386 Supplier Novus Biologicals Supplier No. NB10093386
Add to Cart
Add to Cart

Goat Polyclonal Antibody

COL4A3BP Polyclonal specifically detects COL4A3BP in Mouse samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen COL4A3BP
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1 ug/mL, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_005704.1
Gene Alias Ceramide transfer protein, CERTFLJ20597, CERTL, collagen type IV alpha-3-binding protein, collagen, type IV, alpha 3 (Goodpasture antigen) binding protein, Goodpasture antigen-binding protein, GPBPSTARD11ceramide transporter, hCERT, lipid-transfer protein CERTL, StARD11, StAR-related lipid transfer (START) domain containing 11, StAR-related lipid transfer protein 11, START domain containing 11, START domain-containing protein 11
Gene Symbols CERT1
Host Species Goat
Immunogen Peptide with sequence DHDSAPLNNRC corresponding to internal region according to NP_005704.1, NP_112729.1 .
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cytoskeleton Markers, Immune System Diseases, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10087
Test Specificity This antibody is expected to recognize both reported isoforms (NP_005704.1 and NP_112729.1)
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.