Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL7A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139764
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5139764 100 μL
1 options
This item is not returnable. View return policy

Catalog No. PIPA5139764

Supplier: Invitrogen™ PA5139764

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LTLYTLLEGH EVATPATVVP TGPELPVSPV TDLQATELPG QRVRVSWSPV PGATQYRIIV RSTQGVERTL VLPGSQTAFD LDDVQAGLSY TVRVSARVGP REGSASVLTV RREPETPLAV PGLRVVVSDA TRVRVAWGPA PGASGFRI Highest antigen sequence indentity to the following orthologs: Mouse - 81%, Rat - 76%.

This gene encodes the alpha chain of type VII collagen. The type VII collagen fibril, composed of three identical alpha collagen chains, is restricted to the basement zone beneath stratified squamous epithelia. It functions as an anchoring fibril between the external epithelia and the underlying stroma. Mutations in this gene are associated with all forms of dystrophic epidermolysis bullosa. In the absence of mutations, however, an acquired form of this disease can result from an autoimmune response made to type VII collagen. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL7A1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COL7A1
Gene Accession No. Q02388
Gene Alias alpha-1 type VII collagen; AW209154; COL7A1; collagen alpha-1(VII) chain; collagen type VII alpha 1; collagen type VII alpha 1 chain; collagen vii; collagen VII, alpha-1 polypeptide; collagen, type VII, alpha 1; EBD1; EBDCT; EBR1; HGNC:2214; LC collagen; long chain collagen; long-chain collagen; NDNC8; procollagen, type VII, alpha 1
Gene Symbols COL7A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COL7A1. Recombinant protein control fragment (Product #RP-102926).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1294
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.