Learn More
Description
Specifications
Specifications
| Antigen | COL9A1 |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Dilution | ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | alpha-1 polypeptide, collagen, type IX, alpha 1 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant COL9A1 (P20849-3; Met1-Pro328). |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
