Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COLEC10 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
$178.40 - $511.50
Specifications
| Antigen | COLEC10 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIPA5122078
|
Invitrogen™
PA5122078 |
20 μL |
Each for $178.40
|
|
|||||
PIPA5122077
![]() |
Invitrogen™
PA5122077 |
100 μL |
Each for $511.50
|
|
|||||
Description
COLEC10 is a protein coding gene.Specifications
| COLEC10 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9Y6Z7 | |
| 10584 | |
| Pro34-Lys277 (Accession Q9Y6Z7), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| COLEC10 |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| CL-34; CLL1; CL-L1; Colec10; collectin liver 1; collectin liver protein 1; collectin subfamily member 10; collectin sub-family member 10; collectin sub-family member 10 (C-type lectin); collectin-10; Collectin-34; collectin-L1; UNQ366/PRO702 | |
| COLEC10 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title