Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COLEC10 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988218
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8218 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8218 Supplier Bioss Supplier No. BS8033R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

COLEC10 is a protein coding gene.

Specifications

Antigen COLEC10
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene COLEC10
Gene Accession No. Q9Y6Z7
Gene Alias BOS_13958; CL-34; Cll1; CL-L1; Colec10; collectin liver 1; Collectin liver protein 1; collectin subfamily member 10; collectin sub-family member 10; collectin sub-family member 10 (C-type lectin); collectin sub-family member 10-like; Collectin-10; Collectin-34; collectin-L1; LOC100351735; UNQ366/PRO702
Gene Symbols COLEC10
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human COLEC10.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10584
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.