Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen I alpha 1 Antibody (4), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP221804R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221804R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221804R Supplier Novus Biologicals Supplier No. NBP221804R

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Collagen I alpha 1 Monoclonal specifically detects Collagen I alpha 1 in Human samples. It is validated for ELISA.

Specifications

Antigen Collagen I alpha 1
Applications ELISA
Classification Monoclonal
Clone 4
Conjugate DyLight 550
Dilution ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Alpha-1 type I collagen, COL-IA1, Collagen 1, Collagen 1 alpha 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, Collagen Type I Alpha 1 Chain, collagen, type I, alpha 1, Collagen1, Collagen-1, EDSARTH1, OI4, pro-alpha-1 collagen type 1, Type I Procollagen Alpha 1 Chain
Gene Symbols COL1A1
Host Species Mouse
Immunogen This Collagen I alpha 1 antibody (04) - BSA Free was raised against collagen I purified from second trimester amniotic fluid by affinity chromatography.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Extracellular Matrix, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 1277
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.