Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen I alpha 1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB338057 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338057 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338057 Supplier Novus Biologicals Supplier No. NBP177457APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Collagen I alpha 1 Polyclonal antibody specifically detects Collagen I alpha 1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot (Negative)

Specifications

Antigen Collagen I alpha 1
Applications Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Conjugate APC
Dilution Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Western Blot (Negative)
Formulation PBS
Gene Alias Alpha-1 type I collagen, COL-IA1, Collagen 1, Collagen 1 alpha 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, Collagen Type I Alpha 1 Chain, collagen, type I, alpha 1, Collagen1, Collagen-1, EDSARTH1, OI4, pro-alpha-1 collagen type 1, Type I Procollagen Alpha 1 Chain
Host Species Rabbit
Immunogen This Collagen I alpha 1 antibody was raised against a synthetic peptide made to an internal portion of the human Collagen I protein (between residues 150-200) [UniProt# P02452]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Extracellular Matrix, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 1277
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.