Learn More
Description
Specifications
Specifications
| Antigen | Collagen I |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 532 |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Fluorophore-linked immunosorbent assay |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | Collagen 1, Collagen I |
| Gene Symbols | COL1A1 |
| Host Species | Rabbit |
| Immunogen | Collagen I from human and bovine placenta (Uniprot: P02452) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
