Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen II Antibody (2), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP105169C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105169C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105169C Supplier Novus Biologicals Supplier No. NBP105169C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Collagen II Monoclonal specifically detects Collagen II in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Collagen II
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2
Conjugate DyLight 650
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Alpha-1 type II collagen, ANFH, AOM, cartilage collagen, chondrocalcin, COL11A3, Collagen 2, collagen alpha-1(II) chain, collagen II, alpha-1 polypeptide, collagen type 2, collagen, type II, alpha 1, collagen, type II, alpha 1 (primary osteoarthritis, spondyloepiphysealdysplasia, congenital), Collagen-2, MGC131516, SEDC
Gene Symbols COL2A1
Host Species Mouse
Immunogen Recombinant expressed PIIANP
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1280
Test Specificity Procollagen type IIA N-terminal propeptide
Target Species Human, Canine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.