Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen II Antibody (5B2.5), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N600844J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N600844J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N600844J646 Supplier Novus Biologicals Supplier No. NB600844JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Collagen II Monoclonal specifically detects Collagen II in Human, Mouse, Rat, Chicken, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Collagen II
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 5B2.5
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Alpha-1 type II collagen, ANFH, AOM, cartilage collagen, chondrocalcin, COL11A3, Collagen 2, collagen alpha-1(II) chain, collagen II, alpha-1 polypeptide, collagen type 2, collagen, type II, alpha 1, collagen, type II, alpha 1 (primary osteoarthritis, spondyloepiphysealdysplasia, congenital), Collagen-2, MGC131516, SEDC
Gene Symbols COL2A1
Host Species Mouse
Immunogen A purified preparation of lathyritic type II collagen from embryonic chicken sternum
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cytoskeleton Markers, Extracellular Matrix, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1280.0
Target Species Human, Mouse, Rat, Chicken, Rabbit
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.