Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen III alpha 1/COL3A1 Antibody (1E7-D7/Col3) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018308 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018308 0.025 mg
NBP105119 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018308 Supplier Novus Biologicals Supplier No. NBP1051190.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 18 publications

Collagen III alpha 1/COL3A1 Monoclonal specifically detects Collagen III alpha 1/COL3A1 in Human, Mouse, Rat, Canine, Kangaroo, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Collagen III alpha 1/COL3A1
Applications Immunohistochemistry, ELISA, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 1E7-D7/Col3
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:800, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:300, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen
Formulation 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide
Gene Alias alpha1 (III) collagen, Collagen 3, collagen alpha-1(III) chain, collagen, fetal, collagen, type III, alpha 1, Collagen-3, EDS4A, Ehlers-Danlos syndrome type IV, autosomal dominant, FLJ34534
Gene Symbols COL3A1
Host Species Mouse
Immunogen Acid-digested pepsin soluble human type III collagen
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cytoskeleton Markers, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 1281
Test Specificity Highly specific for type III collagen. It has been shown to have no cross reactivity with type I and V collagens by ELISA and immunoblotting. There is no evidence for cross-reactivity with other connective tissue proteins (laminin, fibronectin, elastin).
Target Species Canine, Rabbit, Kangaroo, Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.