Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen III alpha 1/COL3A1 Antibody (1E7-D7/Col3), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NBP15119646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15119646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP15119646 Supplier Novus Biologicals Supplier No. NBP105119JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Collagen III alpha 1/COL3A1 Monoclonal specifically detects Collagen III alpha 1/COL3A1 in Human, Mouse, Rat, Canine, Kangaroo, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen CollagenIIIalpha1/COL3A1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1E7-D7/Col3
Conjugate Janelia Fluor 646
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias alpha1 (III) collagen, Collagen 3, collagen alpha-1(III) chain, collagen, fetal, collagen, type III, alpha 1, Collagen-3, EDS4A, Ehlers-Danlos syndrome type IV, autosomal dominant, FLJ34534
Gene Symbols COL3A1
Host Species Mouse
Immunogen This Collagen III alpha 1/COL3A1 Antibody (1E7-D7/Col3) was developed against acid-digested pepsin soluble human type III collagen
Molecular Weight of Antigen 139 kDa
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cancer, Cell Biology, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 1281
Test Specificity Highly specific for type III collagen. It has been shown to have no cross reactivity with type I and V collagens by ELISA and immunoblotting. There is no evidence for cross-reactivity with other connective tissue proteins (laminin, fibronectin, elastin).
Target Species Canine, Human, Kangaroo, Rabbit, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.