Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen III alpha 1/COL3A1 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NBP126548 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP126548 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP126548 Supplier Novus Biologicals Supplier No. NBP126548
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Collagen III alpha 1/COL3A1 Polyclonal specifically detects Collagen III alpha 1/COL3A1 in Human samples. It is validated for ELISA.

Specifications

Antigen Collagen III alpha 1/COL3A1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4mg/mL
Conjugate Biotin
Dilution ELISA 1:1000 - 1:4000
Formulation PBS with Sodium Azide
Gene Alias alpha1 (III) collagen, Collagen 3, collagen alpha-1(III) chain, collagen, fetal, collagen, type III, alpha 1, Collagen-3, EDS4A, Ehlers-Danlos syndrome type IV, autosomal dominant, FLJ34534
Gene Symbols COL3A1
Host Species Goat
Immunogen This Collagen III alpha 1/COL3A1 Antibody was developed by hyperimmunizing goats with human type III collagen.
Molecular Weight of Antigen 139 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 1281
Test Specificity Reacts with conformational determinants on human type III collagen as demonstrated by ELISA. May react with type III collagen from other species. Exhibits <10% cross reactivity with collagen type I, II, IV, V and VI. The antibody has not been tested for reactivity with other ECM proteins (e.g., laminin, fibronectin).
Target Species Human, Mouse, Rat, Bovine, Canine
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.