Learn More
Description
Specifications
Specifications
| Antigen | Collagen III alpha 1/COL3A1 |
| Applications | ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:10000, Simple Western, Flow Cytometry, ELISA 1:5000-1:50000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide |
| Gene Accession No. | P02461 |
| Gene Alias | alpha1 (III) collagen, Collagen 3, collagen alpha-1(III) chain, collagen, fetal, collagen, type III, alpha 1, Collagen-3, EDS4A, Ehlers-Danlos syndrome type IV, autosomal dominant, FLJ34534 |
| Gene Symbols | COL3A1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
