Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen IV alpha 1 Antibody (577222), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB003191 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003191 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003191 Supplier Novus Biologicals Supplier No. MAB63081C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Collagen IV alpha 1 Monoclonal specifically detects Collagen IV alpha 1 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen Collagen IV alpha 1
Applications Immunocytochemistry
Classification Monoclonal
Clone 577222
Conjugate DyLight 650
Dilution Immunocytochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias alpha-1 chain, arresten, COL4A1 NC1 domain, collagen alpha-1(IV) chain, collagen IV, alpha-1 polypeptide, collagen, type IV, alpha 1, Collagen-4, EC 3.4.23, EC 6.3.1.2
Gene Symbols COL4A1
Host Species Mouse
Immunogen Partial recombinant human Collagen IV alpha 1 protein (amino acids 1441-1669) [UniProt P02462].
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Extracellular Matrix, Stem Cell Markers
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.