Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen IV Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NBP126550 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP126550 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP126550 Supplier Novus Biologicals Supplier No. NBP126550
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Collagen IV Polyclonal specifically detects Collagen IV in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot.

Specifications

Antigen Collagen IV alpha 1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4mg/mL
Conjugate Biotin
Dilution Western Blot 1:100-1:2000, ELISA 1:1000-1:4000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:20, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:20-1:40, Dot Blot 1:100-1:2000, Immunocytochemistry
Formulation PBS with Sodium Azide
Gene Alias alpha-1 chain, arresten, COL4A1 NC1 domain, collagen alpha-1(IV) chain, collagen IV, alpha-1 polypeptide, collagen, type IV, alpha 1, Collagen-4, EC 3.4.23, EC 6.3.1.2
Gene Symbols COL4A1
Host Species Goat
Immunogen Goats were hyperimmunized with human type IV collagen.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1282
Test Specificity Reacts with conformational determinants on human type IV collagen as demonstrated by ELISA. May react with type IV collagen from other species. Exhibits <10% cross reactivity with collagen type I, II, III, V and VI. The antibody has not been tested for reactivity with other ECM proteins (e.g., laminin, fibronectin).
Target Species Human, Rat, Monkey, Monkey
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.