Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen IV Antibody - BSA Free
SDP

Catalog No. NB11059981 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11059981 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11059981 Supplier Novus Biologicals Supplier No. NB11059981

Rabbit Polyclonal Antibody has been used in 4 publications

Collagen IV Polyclonal specifically detects Collagen IV in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Collagen IV alpha 1
Applications ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:400, Immunohistochemistry 1:100 - 1:400, Immunocytochemistry/Immunofluorescence 1:40-1:60, Immunohistochemistry-Paraffin 1:250-1:1000, Immunohistochemistry-Frozen 1:500
Formulation PBS with 0.09% Sodium Azide
Gene Accession No. P02463
Gene Alias alpha-1 chain, arresten, COL4A1 NC1 domain, collagen alpha-1(IV) chain, collagen IV, alpha-1 polypeptide, collagen, type IV, alpha 1, Collagen-4, EC 3.4.23, EC 6.3.1.2
Gene Symbols COL4A1
Host Species Rabbit
Immunogen Collagen IV purified from mouse EHS tumour
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers, Extracellular Matrix, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1282
Test Specificity NB110-59981 is specific for mouse collagen type IV. The following cross reactivities have been observed: Mouse Type IV (100%), Mouse Type III (<0.1%), Mouse Type II (<0.1%), Mouse Type I (<0.1%), Human Type IV (<0.1%), Human Type V (<0.1%), Mouse fibronectin (<0.1%), Mouse laminin (<0.1%).
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.