Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen IV Antibody (COL-94), Novus Biologicals™
SDP

Catalog No. NB018464 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018464 0.1 mL
NBP122632 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018464 Supplier Novus Biologicals Supplier No. NBP1226320.1ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Collagen IV Monoclonal specifically detects Collagen IV in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Collagen IV alpha 1
Applications Immunohistochemistry
Classification Monoclonal
Clone COL-94
Conjugate Unconjugated
Dilution Immunohistochemistry
Formulation Ascites, containing less than 0.1% sodium azide with Sodium Azide
Gene Alias alpha-1 chain, arresten, COL4A1 NC1 domain, collagen alpha-1(IV) chain, collagen IV, alpha-1 polypeptide, collagen, type IV, alpha 1, Collagen-4, EC 3.4.23, EC 6.3.1.2
Gene Symbols COL4A1
Host Species Mouse
Immunogen Human collagen type IV.
Purification Method Ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1282
Test Specificity This antibody recognizes collagen type IV epitope located on the alpha1 and/or alpha 2 chains of human collagen type IV. This shows no cross reactivity with collagen types I, II, III, V, VI and VII and does not react with human vitronectin, fibronectin or chondroitin sulfate A, B and C.
Target Species Human
Content And Storage Store at 4°C.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.