Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen IV Antibody (COL-94), Novus Biologicals™

Mouse Monoclonal Antibody
$207.00 - $622.00
Specifications
Antigen | Collagen IV alpha 1 |
---|---|
Clone | COL-94 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Ascites |
Description
Collagen IV Monoclonal specifically detects Collagen IV in Human samples. It is validated for Immunohistochemistry.Specifications
Collagen IV alpha 1 | |
Monoclonal | |
Ascites | |
RUO | |
Human | |
1282 | |
Human collagen type IV. | |
Primary | |
Store at 4°C. |
COL-94 | |
Unconjugated | |
Mouse | |
Stem Cell Markers | |
alpha-1 chain, arresten, COL4A1 NC1 domain, collagen alpha-1(IV) chain, collagen IV, alpha-1 polypeptide, collagen, type IV, alpha 1, Collagen-4, EC 3.4.23, EC 6.3.1.2 | |
COL4A1 | |
IgG1 | |
Ascites | |
This antibody recognizes collagen type IV epitope located on the alpha1 and/or alpha 2 chains of human collagen type IV. This shows no cross reactivity with collagen types I, II, III, V, VI and VII and does not react with human vitronectin, fibronectin or chondroitin sulfate A, B and C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title