Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen IV Antibody (M3F7), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB380144 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB380144 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB380144 Supplier Novus Biologicals Supplier No. NBP308515AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Collagen IV Monoclonal antibody specifically detects Collagen IV in Human, Rat, Avian samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Collagen IV
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone M3F7
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias BSVD;BSVD1;COL4A1s;Collagen alpha-1(IV) chain;PADMAL;RATOR
Host Species Mouse
Immunogen alpha1 (IV)-alpha2(IV) triple helix. The epitope is triple helical domain about 900 from the N-terminus
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Extracellular Matrix, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1282
Target Species Human, Rat, Avian
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.