Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen V Antibody, Novus Biologicals™
SDP

Catalog No. NBP126551 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP126551 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP126551 Supplier Novus Biologicals Supplier No. NBP126551
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Collagen V Polyclonal specifically detects Collagen V in Human, Bovine, Equine samples. It is validated for ELISA, Immunohistochemistry-Frozen.

Specifications

Antigen Collagen V
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:1000-1:4000, Immunohistochemistry-Frozen 1:20-1:40
Formulation 0.1M BBS (pH 8.2) with No Preservative
Gene Accession No. P20908
Gene Alias alpha 1 type V collagen, Collagen 5, collagen alpha-1(V) chain, collagen, type V, alpha 1, Collagen-5, EC 2.7.7.6, EC 6.1.1
Gene Symbols COL5A1
Host Species Goat
Immunogen Goats were hyperimmunized with human type V collagen.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cytoskeleton Markers, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 1289
Test Specificity Reacts with conformational determinants on human type V collagen as demonstrated by ELISA. May react with type V collagen from other species. Exhibits <10% cross reactivity with collagen type I, II, III, IV and VI. The antibody has not been tested for reactivity with other ECM proteins (e.g., laminin, fibronectin).
Target Species Human, Bovine, Equine
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.