Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen VI alpha 1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB174614 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB174614 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB174614 Supplier Novus Biologicals Supplier No. NB1206588AF750
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Collagen VI alpha 1 Polyclonal antibody specifically detects Collagen VI alpha 1 in Human, Mouse, Rat, Bovine, Feline samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)

Specifications

Antigen Collagen VI alpha 1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias Collagen 6, collagen alpha-1(VI) chain, collagen VI, alpha-1 polypeptide, collagen, type VI, alpha 1, Collagen-6, human mRNA for collagen VI alpha-1 C-terminal globular domain10alpha 1 (VI) chain (61 AA), OPLL
Host Species Rabbit
Immunogen Collagen VI alpha 1 from human and bovine placenta (Uniprot: P12109)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 1291
Target Species Human, Mouse, Rat, Bovine, Feline
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.