Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen VI alpha 1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP215947 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215947 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215947 Supplier Novus Biologicals Supplier No. NBP215947
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Collagen VI alpha 1 Polyclonal specifically detects Collagen VI alpha 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Collagen VI
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris-Glycine (pH7), 10% Glycerol with 0.01% Thimerosal
Gene Accession No. P12109
Gene Alias Collagen 6, collagen alpha-1(VI) chain, collagen VI, alpha-1 polypeptide, collagen, type VI, alpha 1, Collagen-6, human mRNA for collagen VI alpha-1 C-terminal globular domain10alpha 1 (VI) chain (61 AA), OPLL
Gene Symbols COL6A1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human COL6A1. The exact sequence is proprietary.
Molecular Weight of Antigen 109 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 1291
Test Specificity Human, Mouse. Expected to cross react based on sequence identity: Bovine (87%).
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.