Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen VI alpha 3 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB322116 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB322116 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB322116 Supplier Novus Biologicals Supplier No. NBP297126APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Collagen VI alpha 3 Polyclonal antibody specifically detects Collagen VI alpha 3 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Collagen VI alpha 3
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate APC
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias Collagen Alpha-3(VI) Chain, Collagen VI, Alpha-3 Polypeptide, collagen, type VI, alpha 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Collagen VI alpha 3 (Accession#: P12111-1; 3101T-3177T)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1293
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.