Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen X alpha 1 Antibody (JF0961), Novus Biologicals™
SDP

Catalog No. NBP266988 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266988 Supplier Novus Biologicals Supplier No. NBP266988
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Collagen X alpha 1 Monoclonal specifically detects Collagen X alpha 1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen CollagenXalpha1
Applications Western Blot
Classification Monoclonal
Clone JF0961
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias COL10A1, Collagen Alpha-1(X) Chain, Collagen X, Alpha-1 Polypeptide, Schmid Metaphyseal Chondrodysplasia
Host Species Rabbit
Immunogen Synthetic peptide within Human Collagen X alpha 1 aa 550-599/ 680. (SwissProt: Q03692 Human; SwissProt: Q05306 Mouse; Unigene:20579 Rat)
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 1300
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.