Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen X alpha 1 Rabbit anti-Mouse, Rat, Clone: 0F5Q9, Novus Biologicals™
SDP

Catalog No. NB165934 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165934 100 μg
NB165935 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB165934 Supplier Novus Biologicals Supplier No. NBP315419100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Collagen X alpha 1 Monoclonal antibody specifically detects Collagen X alpha 1 in Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Collagen X alpha 1
Applications Western Blot
Classification Monoclonal
Clone 0F5Q9
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias COL10A1, Collagen Alpha-1(X) Chain, Collagen X, Alpha-1 Polypeptide, Schmid Metaphyseal Chondrodysplasia
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 500-600 of human Collagen X alpha 1 (Q03692).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1300
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.