Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen XI alpha 2 Antibody (473), Novus Biologicals™
SDP

Catalog No. NBP243728 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP243728 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP243728 Supplier Novus Biologicals Supplier No. NBP243728
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence

Collagen XI alpha 2 Monoclonal specifically detects Collagen XI alpha 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Collagen XI alpha 2
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 473
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation PBS with No Preservative
Gene Alias collagen alpha-2(XI) chain, collagen, type XI, alpha 2, DFNA13, DFNB53, HKE5, PARP, pro-a2 chain of collagen type XI, STL3
Gene Symbols COL11A2
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human COL11A2. The exact sequence is proprietary.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1302
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.