Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen XXV alpha 1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB344580 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344580 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344580 Supplier Novus Biologicals Supplier No. NB300249MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Collagen XXV alpha 1 Polyclonal antibody specifically detects Collagen XXV alpha 1 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen Collagen XXV alpha 1
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Alzheimer disease amyloid-associated protein, AMY, CLAC, CLACP, CLAC-P, Collagen 25, collagen alpha-1(XXV) chain, collagen, type XXV, alpha 1, Collagen-25, collagen-like Alzheimer amyloid plaque component, collagenous Alzheimer amyloid plaque component
Host Species Rabbit
Immunogen A synthetic peptide mapping to residues 171-183 [NHGFLSADQQLIK] of the NC2-1 region of human and mouse CLAC-P.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 84570
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.