Learn More
MP Biomedicals™ Collagenase Type I from Clostridium histolyticum
Description
The collagenase product is a mixture of enzymes secreted by C. histolyticum, with different products differentiated by the relative ratios of the 10-18 components found in the secreted enzymes. The main components are two collagenases, clostripain, and a neutral protease. The synergistic action of these enzymes degrade collagen and other intracellular material. The action of both collagenase enzymes and the neutral protease is necessary for effective release of cells from tissue. Various types of collagen are the natural substrates for collagenase.
Unit Definition: One unit will liberate peptides from collagen equivalent in ninhydrin color to 1.0μmol of L-leucine in 5 hr. at pH 7.4 and 37°C in presence of calcium ions. Contains clostripain, neutral protease and trypsin activities.

Specifications
Specifications
Concentration | >125 U/mg |
Color | Brown |
For Use With (Application) | Preparation of isolated cell suspensions |
Content And Storage | Store at 4°C. Desiccate. |
Quantity | 1 g |
Product Type | Collagenase Type I |
Form | Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.