Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Colloidal Silicon Dioxide, NF, EP, 99-100.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
100 g
500 g
5 kg
10 kg
Packaging:
Fiber Drum
Plastic Bottle
Poly Pail
Description
Spectrum™ Chemical Colloidal Silicon Dioxide, NF, EP is used both in the food and pharmaceutical industries as a powder flow helper in the production of tablets. All Spectrum Chemical NF, EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Material | Silicon Dioxide |
| Quantity | 500 g |
| Loss on Drying | 2.5% |
| Loss on Ignition | 5% |
| Packaging | Poly Pail |
| Percent Purity | 99 to 100.5% |
| Grade | Multi-Compendial |
| Chemical Name or Material | Colloidal Silicon Dioxide |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction