Learn More
Novus Biologicals™ Colon Matched Tumor and Normal Tissue Lysate
SDP
Catalog No. NB002094
Click to view available options
Quantity:
2 Vials
Two matched 1 mg/mL vials (100 μg/vial) of human colon tissue lysate, one containing normal tissue and the other tumor tissue, for use in research. Lysates are guaranteed for 6 months from date of receipt.
The Colon Matched Tumor and Normal Tissue Lysate is derived from human.
Specifications
Quantity 2 Vials
Content And Storage Store at -80C. Avoid freeze-thaw cycles.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.