Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ ColorTag Marking Rings For 73 mm
Shop All Eppendorf North America Products
Click to view available options
Color:
Light Yellow
Description
- Flexible silicone rings for easy pipette and dispenser identification and labelling 73 mm inner diameter
- Ideal fit for Eppendorf 12 or 24 channel pipettes (lower part)
- Prevent errors from pipette misidentification and add color and joy to your lab (12 vibrant colors)
- Autoclavable and suitable for UV or H2O2 sterilization as well as extensively tested for lab use
- Unmistakably Eppendorf ColorTag rings have the Eppendorf logo embossed inside for easy recognition of the original
- Eco-friendly, re-closable packaging made from 100 % paper
Specifications
Specifications
| Color | Light Yellow |
| Type | Pipette Lower Part |
| Diameter (Metric) Opening | 73 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction