Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Columbia Agar Base Medium

Catalog No. p-7000074
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Obtain fast and accurate results with a dehydrated culture medium designed for growth of a wide variety of fastidious microorganisms.

Cultivate and isolate a wide variety of fastidious microorganisms using Thermo Scientific™ Columbia Agar Base.

Traditionally, blood agar bases have been either casein hydrolysate or meat infusion media. The advantage of the first lies in the rapid production of large colonies and of the second in clearly defined zones of hemolysis and good colonial differentiation. Columbia Agar Base combines the virtues of both of these types of media to give an improved all-round performance, showing versatility and superior performance in several applications.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Color Straw
Content And Storage 20°C to 25°C
Description Columbia Agar Base Medium
Form Powder
Manufacturing Origin United States
Product Type Agar
Fill Volume 500 g
Quantity 500 g

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.