Learn More
Liofilchem™ ComASP Cefiderocol
SDP
Catalog No. 22778086 Shop All Liofilchem Products
Change view
Click to view available options
Quantity:
4 Systems (panels)
Catalog No. Quantity
22-778-086 4 Systems (panels)
1 options
Non-hazardous according to current legislation but fall within the specific field of application where a safety data sheet must be supplied because they can cause phenomena of sensitisation in sensitive subjects if they come into contact with the skin.
Catalog No. 22-778-086 Supplier Liofilchem™ Supplier No. 75009U
Add to Cart
Add to Cart
Non-hazardous according to current legislation but fall within the specific field of application where a safety data sheet must be supplied because they can cause phenomena of sensitisation in sensitive subjects if they come into contact with the skin.
This is a quantitative broth microdilution method intended for the in vitro determination of antimicrobial susceptibility of bacteria.
  • 4 Panels of ComASP™ Cefiderocol 0.008-128 (2 isolates can be tested on each panel; panels individually packed in foil with silica gel desiccant)
  • 8 Tubes of Iron-Depleted Cation-Adjusted Mueller Hinton Broth
  • Instructions Sheet (includes Test Results Form), available from www.liofilchem.com
Specifications
Antimicrobial Agents Cefiderocol
Concentration 0.008 to 128 μg/mL
Content And Storage Store ComASP™ Cefiderocol 0.008 to 128 at 2°C to 8°C in the original packaging. Do not use the panels beyond the expiry date indicated on the label. Eliminate without using it if there are signs of deterioration.
Product Type Panel
Sterility Non-sterile
Test Type Media Testing
Quantity 4 Systems (panels)