Learn More
Liofilchem™ ComASP Oritavancin
SDP
Catalog No. 22778093 Shop All Liofilchem Products
Change view
Click to view available options
Quantity:
4 Systems (panels)
Catalog No. Quantity
22-778-093 4 Systems (panels)
1 options
Non-hazardous according to current legislation but fall within the specific field of application where a safety data sheet must be supplied because they can cause phenomena of sensitisation in sensitive subjects if they come into contact with the skin.
Catalog No. 22-778-093 Supplier Liofilchem™ Supplier No. 75010
Add to Cart
Add to Cart
Non-hazardous according to current legislation but fall within the specific field of application where a safety data sheet must be supplied because they can cause phenomena of sensitisation in sensitive subjects if they come into contact with the skin.
This system is a compact version of the broth microdilution (BMD) reference method allowing to perform the antimicrobial susceptibility testing of Oritavancin as recommended by international standards (i.e. CLSI, EUCAST, ISO) but in a simpler and less time-consuming way.
  • 4 Systems (panels) of ComASP™ Oritavancin 0.001-16 (panels individually packed in foil with silica gel desiccant)
  • 8 Tubes of Mueller Hinton II Broth w P80 (3.6 mL)
  • Sealing Film
Specifications
Antimicrobial Agents Oritavancin
Concentration 0.001 to 16 μg/mL
Content And Storage Store ComASP™ Oritavancin 0.001 to 16 at 2°C to 8°C in the original packaging. Once an envelope is opened the panel should be used within 7 days and stored at 2°C to 8°C. Keep away from direct sunlight and direct heat
Product Type Panel
Sterility Non-sterile
Test Type Media Testing
Quantity 4 Systems (panels)