Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COMMD1 Antibody (OTI1F2), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP272419A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272419A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272419A30 Supplier Novus Biologicals Supplier No. NBP272419AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

COMMD1 Monoclonal specifically detects COMMD1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen COMMD1
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI1F2
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias C2orf5, COMM domain-containing protein 1, copper metabolism (Murr1) domain containing 1, copper metabolism gene MURR1, MGC27155, MURR1chromosome 2 open reading frame 5 (MURR1), Protein Murr1
Gene Symbols COMMD1
Host Species Mouse
Immunogen Full length recombinant protein of human COMMD1 (NP_689729) produced in HEK293T cells.
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 150684
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.