Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COMMD7 Antibody (49B5), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB309010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309010 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309010 Supplier Novus Biologicals Supplier No. NBP259423MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

COMMD7 Monoclonal antibody specifically detects COMMD7 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen COMMD7
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 49B5
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias C20orf92, chromosome 20 open reading frame 92, COMM domain containing 7, COMM domain-containing protein 7, dJ1085F17.3, FLJ14987, MGC33315
Host Species Mouse
Immunogen Recombinant human COMMD7(1-200aa)purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 149951
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.