Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COMMD8 Antibody (006), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB071970 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB071970 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB071970 Supplier Novus Biologicals Supplier No. NBP290299AF532
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

COMMD8 Monoclonal specifically detects COMMD8 in Human samples. It is validated for ELISA.

Specifications

Antigen COMMD8
Applications ELISA
Classification Monoclonal
Clone 6
Conjugate Alexa Fluor 532
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias COMM domain containing 8, COMM domain-containing protein 8, FLJ20502
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human COMMD8 (AAH19826.1; Met1-Lys183).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54951
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.