Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Common beta Chain Antibody (001), PE, Novus Biologicals™
SDP

Catalog No. NB307133 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB307133 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB307133 Supplier Novus Biologicals Supplier No. NBP289545PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Common beta Chain Monoclonal antibody specifically detects Common beta Chain in Human samples. It is validated for ELISA

Specifications

Antigen Common beta Chain
Applications ELISA
Classification Monoclonal
Clone 001
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias beta, low-affinity, CD131, CD131 antigen, CDw131, colony stimulating factor 2 receptor, beta, low-affinity(granulocyte-macrophage), cytokine receptor common subunit beta, GM-CSF/IL-3/IL-5 receptor common beta subunit, GM-CSF/IL-3/IL-5 receptor common beta-chain, IL3RB, interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3receptor, beta (high affinity)
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Common beta Chain (Accession#: NP_000386.1; Met 1-Trp 443).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1439
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.