Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Common beta Chain Antibody (7B6), Novus Biologicals™
SDP

Catalog No. NB297690 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297690 50 μL
NB297691 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297690 Supplier Novus Biologicals Supplier No. NBP32664350UL
Add to Cart
Add to Cart

Human Monoclonal Antibody

Common beta Chain Monoclonal antibody specifically detects Common beta Chain in Human samples. It is validated for ELISA

Specifications

Antigen Common beta Chain
Applications ELISA
Classification Monoclonal
Clone 7B6
Conjugate Unconjugated
Dilution ELISA
Formulation 50% Glycerol, 0.01M PBS, PH 7.4
Gene Alias beta, low-affinity, CD131, CD131 antigen, CDw131, colony stimulating factor 2 receptor, beta, low-affinity(granulocyte-macrophage), cytokine receptor common subunit beta, GM-CSF/IL-3/IL-5 receptor common beta subunit, GM-CSF/IL-3/IL-5 receptor common beta-chain, IL3RB, interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3receptor, beta (high affinity)
Host Species Human
Immunogen Recombinant Human Common beta Chain protein [UniProt P32927]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1439
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.