Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Common gamma Chain/IL-2 R gamma Antibody (004), DyLight 755
SDP

Catalog No. NB073130 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB073130 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB073130 Supplier Novus Biologicals Supplier No. NBP290406IR

Rabbit Monoclonal Antibody

Common gamma Chain/IL-2 R gamma Monoclonal specifically detects Common gamma Chain/IL-2 R gamma in Mouse samples. It is validated for ELISA.

Specifications

Antigen Common gamma Chain/IL-2 R gamma
Applications ELISA
Classification Monoclonal
Clone 4
Conjugate DyLight 755
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias CD132, CD132 antigen, CIDX, combined immunodeficiency, X-linked, common cytokine receptor gamma chain, common gamma-chain, cytokine receptor common subunit gamma, gamma(c), IL-2 receptor subunit gamma, IL-2R subunit gamma, IL-2RG, IMD4, interleukin 2 receptor, gamma, Interleukin-2 receptor subunit gamma, p64, SCIDX, SCIDX1, severe combined immunodeficiency
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Common gamma Chain/IL-2 R gamma (NP_038591.1; Met1-Ala263).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3561
Test Specificity Has cross-reactivity in ELISA with: Human IL2RG
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.