Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Common gamma Chain/IL-2 R gamma Antibody (633162), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB364014 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB364014 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB364014 Supplier Novus Biologicals Supplier No. FAB2842L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Common gamma Chain/IL-2 R gamma Monoclonal specifically detects Common gamma Chain/IL-2 R gamma in Human samples. It is validated for CyTOF, Flow Cytometry, Neutralization.

Specifications

Antigen Common gamma Chain/IL-2 R gamma
Applications CyTOF, Flow Cytometry, Neutralization
Classification Monoclonal
Clone 633162
Conjugate DyLight 550
Dilution Flow Cytometry, Neutralization, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD132, CD132 antigen, CIDX, combined immunodeficiency, X-linked, common cytokine receptor gamma chain, common gamma-chain, cytokine receptor common subunit gamma, gamma(c), IL-2 receptor subunit gamma, IL-2R subunit gamma, IL-2RG, IMD4, interleukin 2 receptor, gamma, Interleukin-2 receptor subunit gamma, p64, SCIDX, SCIDX1, severe combined immunodeficiency
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf21-derived recombinant human Common gamma Chain/IL-2 gamma , Leu23-Asn254, Accession # P31785
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3561
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.