Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Common gamma Chain/IL-2 R gamma Rabbit anti-Mouse, Clone: 3000000000000, Novus Biologicals™
SDP

Catalog No. NB153641 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB153641 0.025 mg
NB153642 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB153641 Supplier Novus Biologicals Supplier No. NBP3120380.025MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Common gamma Chain/IL-2 R gamma Monoclonal antibody specifically detects Common gamma Chain/IL-2 R gamma in Mouse samples. It is validated for Flow Cytometry, Immunofluorescence, Immunoprecipitation, Neutralization

Specifications

Antigen Common gamma Chain/IL-2 R gamma
Applications Flow Cytometry, Immunofluorescence, Immunoprecipitation, Neutralization
Classification Monoclonal
Clone 3000000000000
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Block/Neutralize
Formulation PBS
Gene Alias CD132, CD132 antigen, CIDX, combined immunodeficiency, X-linked, common cytokine receptor gamma chain, common gamma-chain, cytokine receptor common subunit gamma, gamma(c), IL-2 receptor subunit gamma, IL-2R subunit gamma, IL-2RG, IMD4, interleukin 2 receptor, gamma, Interleukin-2 receptor subunit gamma, p64, SCIDX, SCIDX1, severe combined immunodeficiency
Host Species Rabbit
Immunogen This antibody was raised by immunising Lewis rats with transfected rat myeloma cell line YB2/0, expressing murine cytoplasmic-tailless IL-2R common gamma-chain (YB2/0-myt).
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3561
Target Species Mouse
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.