Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Common gamma Chain/IL-2 R gamma Rabbit anti-Mouse, Clone: 3000000000000, Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | Common gamma Chain/IL-2 R gamma |
---|---|
Clone | 3000000000000 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Block/Neutralize |
Applications | Flow Cytometry, Immunofluorescence, Immunoprecipitation, Neutralization |
Description
Common gamma Chain/IL-2 R gamma Monoclonal antibody specifically detects Common gamma Chain/IL-2 R gamma in Mouse samples. It is validated for Flow Cytometry, Immunofluorescence, Immunoprecipitation, NeutralizationSpecifications
Common gamma Chain/IL-2 R gamma | |
1 mg/mL | |
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Neutralization | |
Unconjugated | |
RUO | |
Mouse | |
CD132, CD132 antigen, CIDX, combined immunodeficiency, X-linked, common cytokine receptor gamma chain, common gamma-chain, cytokine receptor common subunit gamma, gamma(c), IL-2 receptor subunit gamma, IL-2R subunit gamma, IL-2RG, IMD4, interleukin 2 receptor, gamma, Interleukin-2 receptor subunit gamma, p64, SCIDX, SCIDX1, severe combined immunodeficiency | |
This antibody was raised by immunising Lewis rats with transfected rat myeloma cell line YB2/0, expressing murine cytoplasmic-tailless IL-2R common gamma-chain (YB2/0-myt). | |
Primary | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
3000000000000 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Block/Neutralize | |
Monoclonal | |
Rabbit | |
Cancer | |
PBS | |
3561 | |
IgG κ | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title